LIF432341910 Life Chemicals Inc.

1-amino-2-(3-methoxyphenyl)propan-2-ol

  • MDL number MFCD17219190
  • Cas Number : 133562-40-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart