LIF432341924 Life Chemicals Inc.

5-chloro-2-nitrobenzoyl chloride

  • MDL number MFCD02093797
  • Cas Number : 41994-44-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart