LIF432341948 Life Chemicals Inc.

3-benzyl-3,8-diazabicyclo[3.2.1]octane dihydrochloride

  • MDL number MFCD14279200
  • Cas Number : 93428-54-7
  • Molecular Weight : 275.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart