LIF432341955 Life Chemicals Inc.

(4,5,6,7-tetrahydro-1H-benzimidazol-2-ylmethyl)amine dihydrochloride

  • MDL number MFCD26793326
  • Cas Number : 1448125-97-0
  • Molecular Weight : 224.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart