LIF432341956 Life Chemicals Inc.

2-(piperidin-4-yl)-4,5,6,7-tetrahydro-1H-benzo[d]imidazole dihydrochloride

  • MDL number MFCD26793327
  • Cas Number : 1448128-25-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart