LIF432341962 Life Chemicals Inc.

1H-Benzotriazol-5-ylamine hydrochloride

  • MDL number MFCD02683022
  • Cas Number : 93805-11-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart