LIF432341963 Life Chemicals Inc.

Isoxazol-4-ylamine hydrochloride

  • MDL number MFCD09992881
  • Cas Number : 108511-98-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart