LIF432341967 Life Chemicals Inc.

2-chloro-6-piperazin-1-yl-9H-purine dihydrochloride

  • MDL number MFCD22587946
  • Cas Number : 1351612-64-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart