LIF432341968 Life Chemicals Inc.

3-(6,7-dihydro-5H-pyrrolo[1,2-a]imidazol-3-yl)aniline

  • MDL number MFCD12403706
  • Cas Number : 1216284-09-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart