LIF432341969 Life Chemicals Inc.

2-(5,7-Dioxo-5,7-dihydro-pyrrolo[3,4-b]pyridin-6-yl)-ethanesulfonyl chloride hydrochloride

  • MDL number MFCD22587947
  • Cas Number : 1421457-99-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart