LIF432341970 Life Chemicals Inc.

4,5,6,7-tetrahydropyrazolo[1,5-a]pyridine-2-carboxylic acid

  • MDL number MFCD09056814
  • Cas Number : 307313-03-7
  • Molecular Weight : 166.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart