LIF432341973 Life Chemicals Inc.

2-methylimidazo[1,2-a]pyridine-3-carboxylic acid

  • MDL number MFCD00244615
  • Cas Number : 21801-79-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart