LIF432341976 Life Chemicals Inc.

1-(pyridin-2-yl)propan-2-one

  • MDL number MFCD00092717
  • Molecular Weight : 135.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart