LIF432341977 Life Chemicals Inc.

3-(pyridin-2-yl)-1H-pyrazol-5-amine

  • MDL number MFCD06738755
  • Cas Number : 92352-29-9
  • Molecular Weight : 160.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart