LIF432341982 Life Chemicals Inc.

4-chloro-5-(p-tolyl)thieno[2,3-d]pyrimidine

  • MDL number MFCD01238585
  • Cas Number : 374104-63-9
  • Molecular Weight : 260.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart