LIF432341983 Life Chemicals Inc.

4-(3,4-dimethylphenyl)thiazol-2-amine

  • MDL number MFCD00806191
  • Cas Number : 104296-00-6
  • Molecular Weight : 204.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart