LIF432341988 Life Chemicals Inc.

2-amino-6-benzyl-4,5,6,7-tetrahydrothieno[2,3-c]pyridine-3-carbonitrile

  • MDL number MFCD00425859
  • Cas Number : 24237-37-4
  • Molecular Weight : 269.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart