LIF432341993 Life Chemicals Inc.

methyl 2-amino-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carboxylate

  • MDL number MFCD00625612
  • Cas Number : 184174-80-9
  • Molecular Weight : 197.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart