LIF432342001 Life Chemicals Inc.

dimethyl 5-amino-3-methylthiophene-2,4-dicarboxylate

  • MDL number MFCD00159553
  • Cas Number : 103765-33-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart