LIF432342002 Life Chemicals Inc.

(4-phenylthiazol-2-yl)methanamine

  • MDL number MFCD05221867
  • Cas Number : 90916-45-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart