LIF432342005 Life Chemicals Inc.

2-{2-[(4-methylphenyl)sulfanyl]ethyl}piperidine hydrochloride

  • MDL number MFCD08143955
  • Cas Number : 1094873-20-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart