LIF432342026 Life Chemicals Inc.

(m-tolylsulfonyl)valine

  • MDL number MFCD05866550
  • Cas Number : 1007999-32-7
  • Molecular Weight : 271.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart