LIF432342036 Life Chemicals Inc.

naphthalen-2-ylmethanethiol

  • MDL number MFCD00155255
  • Cas Number : 1076-67-1
  • Molecular Weight : 174.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-naphthylmethanethiol
Inchi Code 1S/C11H10S/c12-8-9-5-6-10-3-1-2-4-11(10)7-9/h1-7,12H,8H2
Inchi Key ZSRQBQCAUQRBPS-UHFFFAOYSA-N

Description

Purity 95%
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart