LIF432342038 Life Chemicals Inc.

N-(4-fluorophenyl)-6-morpholino-9H-purin-2-amine

  • MDL number MFCD22608428
  • Cas Number : 1251578-67-2
  • Molecular Weight : 314.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart