LIF432342041 Life Chemicals Inc.

1,3-dimethyl-1,6-dihydro-2H-pyrrolo[3,4-d]pyrimidine-2,4(3H)-dione

  • MDL number MFCD22613718
  • Cas Number : 1396807-57-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart