LIF432342042 Life Chemicals Inc.

2-(3-aminopropyl)-2,3-dihydro-1H-isoindole-1,3-dione hydrochloride

  • MDL number MFCD00035196
  • Cas Number : 121821-01-0
  • Molecular Weight : 240.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart