LIF432342043 Life Chemicals Inc.

4-(cyclopentyloxy)benzaldehyde

  • MDL number MFCD04133155
  • Cas Number : 164520-98-3
  • Molecular Weight : 190.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart