LIF432342055 Life Chemicals Inc.

6-chloroindoline-2,3-dione

  • MDL number MFCD00086347
  • Cas Number : 6341-92-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart