LIF432342056 Life Chemicals Inc.

4-(thiophen-2-yl)benzene-1-sulfonyl chloride

  • MDL number MFCD11655569
  • Cas Number : 209592-91-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart