LIF432342059 Life Chemicals Inc.

(2,2-diethoxyethyl)benzene

  • MDL number MFCD00129694
  • Cas Number : 6314-97-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart