LIF432342061 Life Chemicals Inc.

5-chloro-2-methylbenzo[d]oxazole

  • MDL number MFCD00022850
  • Cas Number : 19219-99-9
  • Molecular Weight : 167.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart