LIF432342066 Life Chemicals Inc.

methyl(methylimino)(phenyl)-lambda6-sulfanone

  • MDL number MFCD00051932
  • Cas Number : 30004-67-2
  • Molecular Weight : 169.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart