LIF432342254 Life Chemicals Inc.

3-(4-(methylsulfonyl)phenyl)propanoic acid

  • MDL number MFCD01319236
  • Cas Number : 387350-46-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart