LIF432342260 Life Chemicals Inc.

7-methoxy-4-methylquinoline

  • MDL number MFCD11655579

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart