LIF432342263 Life Chemicals Inc.

3-(3-fluoro-4-methylphenyl)propanoic acid

  • MDL number MFCD09025364
  • Cas Number : 881189-62-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart