LIF432342279 Life Chemicals Inc.

3-(4-hydrazinylphenyl)propanoic acid hydrochloride

  • MDL number MFCD12922973
  • Cas Number : 105323-52-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart