LIF432342300 Life Chemicals Inc.

3-{2-methylpyrazolo[1,5-a]pyrimidin-6-yl}propan-1-ol

  • MDL number MFCD18917546
  • Cas Number : 83724-99-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart