LIF432342325 Life Chemicals Inc.

ethyl 3-methyl-1,2,4-thiadiazole-5-carboxylate

  • MDL number MFCD22201205
  • Cas Number : 76162-56-6
  • Molecular Weight : 172.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart