LIF432342328 Life Chemicals Inc.

1-tosylpyrrolidin-3-one

  • MDL number MFCD03659114
  • Cas Number : 73696-28-3
  • Molecular Weight : 239.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-[(4-methylphenyl)sulfonyl]-3-pyrrolidinone
Inchi Code 1S/C11H13NO3S/c1-9-2-4-11(5-3-9)16(14,15)12-7-6-10(13)8-12/h2-5H,6-8H2,1H3
Inchi Key BMJNDUPYCKAYAG-UHFFFAOYSA-N

Description

Purity 95%
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart