LIF432342330 Life Chemicals Inc.

ethyl 3-isopropyl-1H-pyrazole-5-carboxylate

  • MDL number MFCD03232642
  • Cas Number : 78208-72-7
  • Molecular Weight : 182.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart