LIF432342334 Life Chemicals Inc.

3-(piperidin-1-ylmethyl)-1,2,4-thiadiazol-5-amine

  • MDL number MFCD21092110
  • Cas Number : 1352999-69-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart