LIF432342339 Life Chemicals Inc.

3-(pyridin-2-yloxy)benzaldehyde

  • MDL number MFCD03840110
  • Cas Number : 137386-78-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart