LIF432342345 Life Chemicals Inc.

2-((phenylsulfonyl)methyl)pyrrolidine hydrochloride

  • MDL number MFCD12028438
  • Cas Number : 1185302-64-0
  • Molecular Weight : 261.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart