LIF432342348 Life Chemicals Inc.

6,7-dihydro-5H-pyrrolo[3,4-d]pyrimidine dihydrochloride

  • MDL number MFCD08457576
  • Cas Number : 157327-51-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart