LIF432342351 Life Chemicals Inc.

tert-butyl (1-(4-bromothiazol-2-yl)piperidin-4-yl)carbamate

  • MDL number MFCD26954585
  • Cas Number : 1289050-83-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart