LIF432342356 Life Chemicals Inc.

methyl 7-methoxy-4-nitro-1H-indazole-3-carboxylate

  • MDL number MFCD26954588
  • Cas Number : 1448066-97-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart