LIF432342357 Life Chemicals Inc.

methyl 4-amino-7-methoxy-1H-indazole-3-carboxylate

  • MDL number MFCD26954589
  • Cas Number : 1448125-93-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart