LIF432342358 Life Chemicals Inc.

3-(phenylsulfonyl)-8-azabicyclo[3.2.1]octane hydrochloride

  • MDL number MFCD22420556
  • Cas Number : 1135000-80-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart