LIF432342360 Life Chemicals Inc.

4-[(2-furylmethyl)sulfonyl]piperidine hydrochloride

  • MDL number MFCD24842730
  • Cas Number : 1448050-07-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart