LIF432342365 Life Chemicals Inc.

2-(morpholinomethyl)acrylic acid

  • MDL number MFCD06208438
  • Cas Number : 4432-44-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart